ADV390218276 AChemBlock

Edoxaban

  • MDL number MFCD13195544
  • Cas Number : 480449-70-5
  • Molecular Weight : 548.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form white powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/edoxaban.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5828
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart