ADV390218279 AChemBlock

(2,2-Difluoro-benzo[1,3]dioxol-5-yl)-acetic acid

  • MDL number MFCD09925076
  • Cas Number : 398156-38-2
  • Molecular Weight : 216.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-2-difluoro-benzo-1-3-dioxol-5-yl-acetic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4475
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart