ADV390218293 AChemBlock

2-Amino-5-methoxypyrimidine

  • MDL number MFCD00187864
  • Cas Number : 13418-77-4
  • Molecular Weight : 125.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-amino-5-methoxypyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5676
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart