ADV390218304 AChemBlock

4-bromo-1,3-dihydro-2H-inden-2-one

  • MDL number MFCD09744095
  • Cas Number : 846032-36-8
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2h-inden-2-one-4-bromo-1-3-dihydro.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5797
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart