ADV390218306 AChemBlock

Methyl 5,6-dihydroxypyridine-2-carboxylate

  • MDL number MFCD09838989
  • Cas Number : 1260883-63-3
  • Molecular Weight : 169.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-6-dihydroxypyridine-2-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5826
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart