ADV390218314 AChemBlock

3-Boc-6-oxo-3-azabicyclo[3.2.0]heptane

  • MDL number MFCD12911861
  • Cas Number : 663172-80-3
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-boc-6-oxo-3-azabicyclo-3-2-0-heptane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6019
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart