ADV390218327 AChemBlock

5-Bromo-2-chloro-3,4-pyridinediamine

  • MDL number MFCD02266941
  • Cas Number : 163452-78-6
  • Molecular Weight : 222.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-chloro-3-4-pyridinediamine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6034
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart