ADV390218328 AChemBlock

2-Boc-1,2,3,4-tetrahydro-isoquinoline-7-carboxylic acid

  • MDL number MFCD03094736
  • Cas Number : 149353-95-7
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form gray solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-boc-1-2-3-4-tetrahydro-isoquinoline-7-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6036
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart