ADV390218337 AChemBlock

2-Aminomethyl-3-chlorothiophene hydrochloride

  • MDL number MFCD22423096
  • Cas Number : 643088-03-3
  • Molecular Weight : 184.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-aminomethyl-3-chlorothiophene-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6088
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart