ADV390218344 AChemBlock

3-Methyl-5-nitro-1H-indazole

  • MDL number MFCD07082699
  • Cas Number : 40621-84-9
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-5-nitro-1h-indazole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6117
pictograms GHS07
hstatements H303
Availability details for
Quantity
  • Availability
  • Add to Cart