ADV390218345 AChemBlock

3-Oxa-9-azabicyclo[3.3.1]nonan-7-one hydrochloride

  • MDL number MFCD20491410
  • Cas Number : 1126795-00-3
  • Molecular Weight : 177.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-oxa-9-azabicyclo-3-3-1-nonan-7-one-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6367
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart