ADV390218347 AChemBlock

2-Cbz-6-hydroxy-2-azaspiro[3.3]heptane

  • MDL number MFCD22415206
  • Cas Number : 1363383-32-7
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-cbz-6-hydroxy-2-azaspiro-3-3-heptane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6463
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart