ADV390218355 AChemBlock

Methyl 1-methyl-4-nitro-1H-pyrazole-5-carboxylate

  • MDL number MFCD02110822
  • Cas Number : 309740-49-6
  • Molecular Weight : 185.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-methyl-4-nitro-1h-pyrazole-5-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6490
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart