ADV390218364 AChemBlock

exo-8-Boc-3-cyano-8-azabicyclo[3.2.1]octane

  • MDL number MFCD23381617
  • Cas Number : 1147558-40-4
  • Molecular Weight : 236.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/exo-8-boc-3-cyano-8-azabicyclo-3-2-1-octane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6511
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart