ADV390218365 AChemBlock

5-Fluoro-2-thiophenemethamine

  • MDL number MFCD18324169
  • Cas Number : 214759-36-1
  • Molecular Weight : 131.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-fluoro-2-thiophenemethamine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6513
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart