ADV390218369 AChemBlock

2,2-Difluoro-1,3-benzodioxole-5-acetonitrile

  • MDL number MFCD09923942
  • Cas Number : 68119-31-3
  • Molecular Weight : 197.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-2-difluoro-1-3-benzodioxole-5-acetonitrile.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6544
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart