ADV390218374 AChemBlock

7-Boc-7-azaspiro[3.5]nonane-2-carbaldehyde

  • MDL number MFCD20926149
  • Cas Number : 1346556-73-7
  • Molecular Weight : 253.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-boc-7-azaspiro-3-5-nonane-2-carbaldehyde.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5967
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart