ADV390218386 AChemBlock

tert-Butyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydropyridine-1(2H)-carboxylate

  • MDL number MFCD07776601
  • Cas Number : 865245-32-5
  • Molecular Weight : 309.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-6-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-3-4-dihydropyridine-1-2h-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4346
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart