ADV390218391 AChemBlock

5-Nitro-2-(trifluoromethoxy)benzoic acid

  • MDL number MFCD11847469
  • Cas Number : 307989-55-5
  • Molecular Weight : 251.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-nitro-2-trifluoromethoxy-benzoic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4353
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart