ADV390218394 AChemBlock

5-(Trifluoromethyl)-1H-pyrazole

  • MDL number MFCD09860582
  • Cas Number : 20154-03-4
  • Molecular Weight : 136.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-trifluoromethyl-1h-pyrazole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4356
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart