ADV390218408 AChemBlock

1-(2-Chloro-6-fluorophenyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD11036974
  • Cas Number : 1006876-25-0
  • Molecular Weight : 214.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-2-chloro-6-fluorophenyl-cyclopropane-1-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4371
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart