ADV390218413 AChemBlock

3-Cbz-6-oxa-3-aza-bicyclo[3.1.0]hexane

  • MDL number MFCD09608058
  • Cas Number : 31865-25-5
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form light yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-cbz-6-oxa-3-aza-bicyclo-3-1-0-hexane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4376
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart