ADV390218424 AChemBlock

7-Bromo-2,2-dimethylchroman-4-one

  • MDL number MFCD13190381
  • Cas Number : 130200-01-0
  • Molecular Weight : 255.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-2-2-dimethylchroman-4-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4387
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart