ADV390218438 AChemBlock

1-Bromo-4-methoxy-2,3-dinitrobenzene

  • MDL number MFCD20484891
  • Cas Number : 860465-95-8
  • Molecular Weight : 277.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-bromo-4-methoxy-2-3-dinitrobenzene.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4408
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart