ADV390218439 AChemBlock

methyl 3-cyano-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

  • MDL number MFCD18383384
  • Cas Number : 1258963-20-0
  • Molecular Weight : 287.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-cyano-4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-benzoic-acid-methyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4409
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart