ADV390218453 AChemBlock

4,4-Difluoro-pyrrolidine-1,3-dicarboxylic acid 1-tert-butyl ester

  • MDL number MFCD13189575
  • Cas Number : 1196145-11-5
  • Molecular Weight : 251.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-4-difluoro-pyrrolidine-1-3-dicarboxylic-acid-1-tert-butyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4423
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart