ADV390218457 AChemBlock

4-Morpholinocyclohexanone

  • MDL number MFCD17013167
  • Cas Number : 139025-93-7
  • Molecular Weight : 183.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-morpholinocyclohexanone.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4427
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart