ADV390218500 AChemBlock

3,5-Dichloro-2-nitrobenzaldehyde

  • MDL number MFCD01110976
  • Cas Number : 59178-12-0
  • Molecular Weight : 220.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-5-dichloro-2-nitrobenzaldehyde.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4468
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart