ADV390218515 AChemBlock

3-Boc-3,8-diazabicyclo[3.2.1]octane

  • MDL number MFCD17016700
  • Cas Number : 201162-53-0
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form white solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-boc-3-8-diazabicyclo-3-2-1-octane.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4488
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart