ADV390218550 AChemBlock

1-(1-Bromo-2,2,2-trifluoroethyl)-4-methoxybenzene

  • MDL number MFCD12789706
  • Cas Number : 104395-39-3
  • Molecular Weight : 269.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-1-bromo-2-2-2-trifluoroethyl-4-methoxybenzene.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4527
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart