ADV412110164 AChemBlock

2-Boc-2,5-diazaspiro[3.4]octane hemioxalate

  • MDL number MFCD26959054
  • Cas Number : 1359655-69-8
  • Molecular Weight : 514.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-boc-2-5-diazaspiro-3-4-octane-hemioxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4179
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart