ADV412110165 AChemBlock

1-Boc-1,6-diazaspiro[3.3]heptane hemioxalate

  • MDL number MFCD26959055
  • Cas Number : 1523571-10-9
  • Molecular Weight : 486.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-1-6-diazaspiro-3-3-heptane-hemioxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4180
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart