ADV412110172 AChemBlock

1-(2,2,2-Trifluoroethyl)azetidin-3-amine dihydrochloride

  • MDL number MFCD26959070
  • Cas Number : 1820648-37-0
  • Molecular Weight : 227.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-2-2-2-trifluoroethyl-azetidin-3-amine-dihydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4772
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart