ADV412110182 AChemBlock

Exo-(3-oxa-bicyclo[3.1.0]hex-6-yl)-methanol

  • MDL number MFCD27986972
  • Cas Number : 135577-15-0
  • Molecular Weight : 114.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/exo-3-oxa-bicyclo-3-1-0-hex-6-yl-methanol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8001
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart