ADV412110189 AChemBlock

2-Cyanopyrrolidine TFA

  • MDL number MFCD26959084
  • Cas Number : 1523606-34-9
  • Molecular Weight : 210.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-cyanopyrrolidine-tfa.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6826
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart