ADV412110206 AChemBlock

Methyl 5-bromo-1-(tetrahydropyran-2-yl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD26959101
  • Cas Number : 1434142-09-2
  • Molecular Weight : 289.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromo-1-tetrahydropyran-2-yl-1h-pyrazole-4-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6952
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart