ADV412110220 AChemBlock

1-(Azetidin-3-yl)azetidin-3-ol oxalate

  • MDL number MFCD26959117
  • Cas Number : 1523606-25-8
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-azetidin-3-yl-azetidin-3-ol-oxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7004
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart