ADV412110225 AChemBlock

ethyl 7-chloro-3-methyl-1H-pyrrolo[3,2-b]pyridine-2-carboxylate

  • MDL number MFCD26959121
  • Cas Number : 1434141-73-7
  • Molecular Weight : 238.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrrolo-3-2-b-pyridine-2-carboxylic-acid-7-chloro-3-methyl-ethyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7046
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart