ADV412110236 AChemBlock

(3aS,5R,6S,6aS)-5-(((tert-butyldiphenylsilyl)oxy)methyl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol

  • MDL number MFCD26959134
  • Cas Number : 133048-89-2
  • Molecular Weight : 428.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/beta-d-lyxofuranose-5-o-1-1-dimethylethyl-diphenylsilyl-1-2-o-1-methylethylidene.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7109
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart