ADV412110239 AChemBlock

cis-1,3-Cyclobutanediamine dihydrochloride

  • MDL number MFCD26959136
  • Cas Number : 1523571-90-5
  • Molecular Weight : 159.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/cis-1-3-cyclobutanediamine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7228
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart