ADV412110245 AChemBlock

7-Thia-2-aza-spiro[3.5]nonane 7,7-dioxide hemioxalate

  • MDL number MFCD26959144
  • Cas Number : 1501856-47-8
  • Molecular Weight : 440.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-thia-2-aza-spiro-3-5-nonane-7-7-dioxide-hemioxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7275
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart