ADV412110262 AChemBlock

Telaprevir

  • MDL number MFCD11616089
  • Cas Number : 402957-28-2
  • Molecular Weight : 679.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/telaprevir.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4593
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart