ADV412110263 AChemBlock

3-Hydroxy-1-methylazetidine hydrochloride

  • MDL number MFCD16605978
  • Cas Number : 26687-49-0
  • Molecular Weight : 123.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-hydroxy-1-methylazetidine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2454-1
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart