ADV412110279 AChemBlock

2,9-Diazaspiro[5.5]undecan-1-one hydrochloride

  • MDL number MFCD08461145
  • Cas Number : 1187173-73-4
  • Molecular Weight : 204.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-9-diazaspiro-5-5-undecan-1-one-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4614
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart