ADV412110292 AChemBlock

Methyl 4-amino-3,5-difluorobenzoate

  • MDL number MFCD20725083
  • Cas Number : 1162676-00-7
  • Molecular Weight : 187.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-amino-3-5-difluorobenzoate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4629
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart