ADV412110294 AChemBlock

2-Oxa-6-azaspiro[3.3]heptane-6-ethanamine

  • MDL number MFCD13184501
  • Cas Number : 54384-40-6
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-oxa-6-azaspiro-3-3-heptane-6-ethanamine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4631
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart