ADV412110306 AChemBlock

1,1-Dimethyl 3-oxocyclobutane-1,1-dicarboxylate

  • MDL number MFCD21122007
  • Cas Number : 1486409-21-5
  • Molecular Weight : 186.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-1-dimethyl-3-oxocyclobutane-1-1-dicarboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4644
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart