ADV412110329 AChemBlock

(8as)-Octahydro-8a-methyl-pyrrolo[1,2-a]pyrazine

  • MDL number MFCD19227823
  • Cas Number : 1261569-85-0
  • Molecular Weight : 140.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/8as-octahydro-8a-methyl-pyrrolo-1-2-a-pyrazine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4670
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart