ADV412110345 AChemBlock

4-(Methoxycarbonyl)bicyclo[2.2.2]octane-1-carboxylic acid

  • MDL number MFCD18459359
  • Cas Number : 18720-35-9
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-methoxycarbonyl-bicyclo-2-2-2-octane-1-carboxylic-acid.html

Safety Information

signalword Danger
pcodes P264;P270;P301+310+330
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4688
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart