ADV412110355 AChemBlock

2,2-Dimethyltetrahydropyran-4-one

  • MDL number MFCD01549337
  • Cas Number : 1194-16-7
  • Molecular Weight : 128.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-2-dimethyltetrahydropyran-4-one.html
Availability details for
Quantity
  • Availability
  • Add to Cart